(Bis(trifluoroacetoxy)iodo)benzene 97%

Supplier: Thermo Scientific Chemicals
Warning

L15141-06 L15141-14
CAAAAL15141-06EA 72.36 CAD
CAAAAL15141-06 CAAAAL15141-14
(Bis(trifluoroacetoxy)iodo)benzene 97%
(Bis(trifluoroacetoxy)iodo)benzene

Fieser: 9,54 13,241 14,257 15,257 16,274 18,289 19,41 20,305 21,342


Purity: 97% CAS No.: 2712-78-9 Molecular Formula: C10H5F6IO4 Formula Weight: 430.04 Storage Temperature: 2°C to 8°C Melting Point: 122-125° Beilstein Reference: 764767 MDL No.: MFCD00009672

Formula: C₁₀H₅F₆IO₄
Melting Pt: 122…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00009672
CAS Number: 2712-78-9
EINECS: 220-308-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR