5-Methoxyindole-2-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals

L15137-03 L15137-06
CAAAAL15137-03EA 43.61 CAD
CAAAAL15137-03 CAAAAL15137-06
5-Methoxyindole-2-carboxylic acid 97%
5-Methoxyindole-2-carboxylic acid

Purity: 97% CAS No.: 4382-54-1 Molecular Formula: C10H9NO3 Formula Weight: 191.19 Melting Point: ca 200° dec. Beilstein Reference: 157478 MDL No.: MFCD00005614

Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
Melting Pt: ∼ 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005614
CAS Number: 4382-54-1
EINECS: 224-487-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR