Sodium-tert-pentyloxide 95%

Supplier: Thermo Scientific Chemicals
Danger

L15112-14 L15112-22
CAAAAL15112-14EA 58.62 CAD
CAAAAL15112-14 CAAAAL15112-22
Sodium-tert-pentyloxide 95%
Sodium-tert-pentyloxide

Purity: 95% CAS No.: 14593-46-5 Molecular Formula: C5H11NaO Formula Weight: 110.13 Odor: Odorless Beilstein Reference: 3983987 UN No.: 3206 MDL No.: MFCD00064245

Formula: C₅H₁₁NaO
Storage Temperature: Ambient
MDL Number: MFCD00064245
CAS Number: 14593-46-5
EINECS: 238-369-4
UN: 3206
ADR: 4.2,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR