Borane-morpholine complex 97%

Supplier: Thermo Scientific Chemicals
Danger

L15110-06 L15110-14
CAAAAL15110-06EA 57.67 CAD
CAAAAL15110-06 CAAAAL15110-14
Borane-morpholine complex 97%
Borane-morpholine complex

Purity: 97% CAS No.: 4856-95-5 Molecular Formula: C4H12BNO Formula Weight: 100.96 Odor: Amine-like Melting Point: 93-95° UN No.: 2925 MDL No.: MFCD00066736

Formula: C₄H₁₂BNO
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00066736
CAS Number: 4856-95-5
EINECS: 225-450-7
UN: 2925
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR