3-Carboxy-2,2,5,5-tetramethylpyrrolidine-1-oxyl 97 radical_free
Supplier: Thermo Scientific Chemicals
Synonyms:
3-Carboxy-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, 3-Carboxy-PROXYL
L15068-MD
L15068-MF
CAAAAL15068-MDEA
205.4
CAD
CAAAAL15068-MD
CAAAAL15068-MF
3-Carboxy-2,2,5,5-tetramethylpyrrolidine-1-oxyl 97 radical_free
3-Carboxy-2,2,5,5-tetramethylpyrrolidine-1-oxyl
Purity: 97+%
CAS No.: 2154-68-9
Molecular Formula: C9H16NO3
Formula Weight: 186.23
Melting Point: ca 200° dec.
Beilstein Reference: 4136411
MDL No.: MFCD00003167
Formula:
C₉H₁₆NO₃ MW: 186.23 g/mol Melting Pt: ∼ 200 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003167 CAS Number: 2154-68-9 EINECS: 218-448-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...