Sodium bromoacetate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L14983-06
L14983-14
CAAAAL14983-06EA
65.1
CAD
CAAAAL14983-06
CAAAAL14983-14
Sodium bromoacetate 98%
Sodium bromoacetate
Purity: 98%
CAS No.: 1068-52-6
Molecular Formula: BrCH2CO2Na
Formula Weight: 160.93
Odor: Odorless
Melting Point:
Formula:
BrCH₂CO₂Na Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD01311832 CAS Number: 1068-52-6 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...