Ethyl-4-hydroxy-3-methoxycinnamate 98%
Supplier: Thermo Scientific Chemicals
L14960-03
L14960-MD
CAAAAL14960-03EA
130.03
CAD
CAAAAL14960-03
CAAAAL14960-MD
Ethyl-4-hydroxy-3-methoxycinnamate 98%
Ethyl-4-hydroxy-3-methoxycinnamate
Purity: 99%
CAS No.: 4046-02-0
Molecular Formula: C12H14O4
Formula Weight: 222.24
Melting Point: 56-58°
Beilstein Reference: 2696807
MDL No.: MFCD00009190
Formula:
C₁₂H₁₄O₄ Melting Pt: 56…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009190 CAS Number: 4046-02-0 EINECS: 223-745-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...