Arbutin 98+% (by HPLC)
Supplier: Thermo Scientific Chemicals
L14945-03
L14945-06
CAAAAL14945-03EA
49.77
CAD
CAAAAL14945-03
CAAAAL14945-06
Arbutin 98+% (by HPLC)
Arbutin
Purity: 98+%
CAS No.: 497-76-7
Molecular Formula: C12H16O7
Formula Weight: 272.26
Melting Point: 196-200°
Beilstein Reference: 89673
Optical Rotation: -65° (c=2 in water)
MDL No.: MFCD00016915
Formula:
C₁₂H₁₆O₇ Melting Pt: 196…200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016915 CAS Number: 497-76-7 EINECS: 207-850-3 Merck Index: 14,773 |
Specification Test Results
Optical rotation | -65° (c=2 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...