1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L14835-03
L14835-06
CAAAAL14835-03EA
105.86
CAD
CAAAAL14835-03
CAAAAL14835-06
1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine 97%
1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine
Purity: 97%
CAS No.: 202823-67-4
Molecular Formula: C10H10Cl2N2
Formula Weight: 229.11
Melting Point: 208-211°
MDL No.: MFCD01075667
Formula:
C₁₀H₁₀Cl₂N₂ Melting Pt: 208…211 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075667 CAS Number: 202823-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...