4-(Trimethylsilyl)-3-butyn-2-ol 97%

Supplier: Thermo Scientific Chemicals
Warning

L14815-06 L14815-14
CAAAAL14815-06EA 109.34 CAD
CAAAAL14815-06 CAAAAL14815-14
4-(Trimethylsilyl)-3-butyn-2-ol 97%
4-(Trimethylsilyl)-3-butyn-2-ol

Purity: 97% CAS No.: 6999-19-5 Molecular Formula: C7H14OSi Formula Weight: 142.27 Storage Temperature: 2°C to 8°C Flash Point: 57°(135°F) Refractive Index: 1.4460 Boiling Point: 126-129° Beilstein Reference: 1923632 UN No.: 1987 MDL No.: MFCD00190213

Formula: C₇H₁₄OSi
Boiling Pt: 126…129 °C
Density: 0.846
Flash Pt: 57 °C(135 °F)
Storage Temperature: Ambient
MDL Number: MFCD00190213
CAS Number: 6999-19-5
UN: 1987
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR