4-Methoxy-2-methylphenylhydrazine hydrochloride 96%

Supplier: Thermo Scientific Chemicals
Warning

L14810-03 L14810-06
CAAAAL14810-03EA 63.99 CAD
CAAAAL14810-03 CAAAAL14810-06
4-Methoxy-2-methylphenylhydrazine hydrochloride 96%
4-Methoxy-2-methylphenylhydrazine hydrochloride

Purity: 96% CAS No.: 93048-16-9 Molecular Formula: C8H12N2O.HCl Formula Weight: 188.66 Melting Point: ca 110° dec. Beilstein Reference: 7227704 UN No.: 2811 MDL No.: MFCD03426021

Formula: C₈H₁₂N₂O·HCl
Melting Pt: ca 110 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03426021
CAS Number: 93048-16-9
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR