Diethyl (3-chloropropyl)malonate 98%

Supplier: Thermo Scientific Chemicals
Warning

L14776-06 L14776-14
CAAAAL14776-06EA 54.19 CAD
CAAAAL14776-06 CAAAAL14776-14
Diethyl (3-chloropropyl)malonate 98%
Diethyl (3-chloropropyl)malonate

Purity: 98% CAS No.: 18719-43-2 Molecular Formula: C10H17ClO4 Formula Weight: 236.70 Flash Point: 110°(230°F) Refractive Index: 1.4430 Boiling Point: 102°/1mm Beilstein Reference: 1785711 MDL No.: MFCD00009169

Formula: C₁₀H₁₇ClO₄
Boiling Pt: 102 °C/1 mm
Density: 1.101
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009169
CAS Number: 18719-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR