Benzimidazole-5,6-dicarboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L14754-03
L14754-06
CAAAAL14754-03EA
79.95
CAD
CAAAAL14754-03
CAAAAL14754-06
Benzimidazole-5,6-dicarboxylic acid 97%
Benzimidazole-5,6-dicarboxylic acid
Purity: 97%
CAS No.: 10351-75-4
Molecular Formula: C9H6N2O4
Formula Weight: 206.16
Melting Point: ca 295° dec.
Beilstein Reference: 528170
MDL No.: MFCD03093058
Formula:
C₉H₆N₂O₄ Melting Pt: ca 295 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD03093058 CAS Number: 10351-75-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...