trans-1-Methoxy-3-trimethylsilyloxy-1,3-butadiene 96%

Supplier: Thermo Scientific Chemicals
Warning

L14672-03 L14672-06
CAAAAL14672-03EA 116.13 CAD
CAAAAL14672-03 CAAAAL14672-06
trans-1-Methoxy-3-trimethylsilyloxy-1,3-butadiene 96%
trans-1-Methoxy-3-trimethylsilyloxy-1,3-butadiene

Fieser: 6,370 7,233 9,303 10,260 11,332 12,318 16,163 17,182 18,236


Purity: 97% CAS No.: 54125-02-9 Molecular Formula: C8H16O2Si Formula Weight: 172.30 Storage Temperature: 2°C to 8°C Flash Point: 45°(113°F) Refractive Index: 1.4550 Boiling Point: 68-69°/14mm Beilstein Reference: 1616761 UN No.: 1993 MDL No.: MFCD00008498

Formula: C₈H₁₆O₂Si
Boiling Pt: 68…69 °C/14 mm
Density: 0.890
Flash Pt: 45 °C(113 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008498
CAS Number: 54125-02-9
UN: 1993
ADR: "3",III
Merck Index: 14,6001

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR