1-Bromo-3,5-di-tert-butylbenzene 99%
Supplier: Thermo Scientific Chemicals
L14646-03
L14646-06
L14646-14
CAAAAL14646-03EA
75.68
CAD
CAAAAL14646-03
CAAAAL14646-06
CAAAAL14646-14
1-Bromo-3,5-di-tert-butylbenzene 99%
1-Bromo-3,5-di-tert-butylbenzene
Purity: 99%
CAS No.: 22385-77-9
Molecular Formula: C14H21Br
Formula Weight: 269.23
Melting Point: 62-66°
Beilstein Reference: 2091559
Formula:
C₁₄H₂₁Br Melting Pt: 62…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00796945 CAS Number: 22385-77-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...