2,2,7-Trimethyloctane-3,5-dione ≥98%
Supplier: Thermo Scientific Chemicals
L14639-03
L14639-06
CAAAAL14639-03EA
72.21
CAD
CAAAAL14639-03
CAAAAL14639-06
2,2,7-Trimethyloctane-3,5-dione ≥98%
2,2,7-Trimethyloctane-3,5-dione
Purity: 98+%
CAS No.: 69725-37-7
Molecular Formula: C11H20O2
Formula Weight: 184.28
Refractive Index: 1.4580
Boiling Point: 44°/0.2mm
MDL No.: MFCD00511279
Formula:
C₁₁H₂₀O₂ Boiling Pt: 44 °C/0.2 mm Storage Temperature: Ambient |
MDL Number:
MFCD00511279 CAS Number: 69725-37-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...