6-Methoxy-2-naphthoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

L14610-03 L14610-06
CAAAAL14610-03EA 61.62 CAD
CAAAAL14610-03 CAAAAL14610-06
6-Methoxy-2-naphthoic acid 98+%
6-Methoxy-2-naphthoic acid

Purity: 98+% CAS No.: 2471-70-7 Molecular Formula: C12H10O3 Formula Weight: 202.21 Melting Point: 192-194° Beilstein Reference: 2211233

Formula: C₁₂H₁₀O₃
MW: 202.21 g/mol
Melting Pt: 192…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00092750
CAS Number: 2471-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR