m-Tolyl acetate 97%
Supplier: Thermo Scientific Chemicals
L14602-14
L14602-22
CAAAAL14602-14EA
58.93
CAD
CAAAAL14602-14
CAAAAL14602-22
m-Tolyl acetate 97%
m-Tolyl acetate
Purity: 97%
CAS No.: 122-46-3
Molecular Formula: C9H10O2
Formula Weight: 150.18
Flash Point: 95°(203°F)
Refractive Index: 1.5020
Boiling Point: 100°/14mm
Beilstein Reference: 1862796
Merck Reference: 14,2583
MDL No.: MFCD00041910
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Boiling Pt: 100 °C/14 mm Density: 1.050 Flash Pt: 95 °C(203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041910 CAS Number: 122-46-3 EINECS: 204-546-2 Merck Index: 14,2583 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...