(S)-(-)-2-Methyl-CBS-oxazaborolidine 1 M in toluene

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-5,5-Diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, (S)-Me-CBS Catalyst, (S)-3,3-Diphenyl-1-methyltetrahydro-1H,3H-pyrrolo[1,2-c][1,3,2]oxazaborole, (S)-3,3-Diphenyl-1-methylpyrrolidino[1,2-c]-1,3,2-oxazaborole

L14583-AA L14583-AC
CAAAAL14583-AAEA 129.56 CAD
CAAAAL14583-AA CAAAAL14583-AC CAAAAL14583-XF
(S)-(-)-2-Methyl-CBS-oxazaborolidine 1 M in toluene
(S)-(-)-2-Methyl-CBS-oxazaborolidine

Fieser: 14,239 16,253 17,242


CAS No.: 112022-81-8 Molecular Formula: C18H20BNO Formula Weight: 277.17 Flash Point: 4°(39°F) UN No.: 1993 MDL No.: MFCD00078439

Formula: C₁₈H₂₀BNO
MW: 277.17 g/mol
Density: 0.925
Flash Pt: 4 °C (39 °F)
Storage Temperature: Ambient
MDL Number: MFCD00078439
CAS Number: 112022-81-8
UN: 1294
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR