(R)-(+)-2-Methyl-CBS-oxazaborolidine 1 M in toluene
Supplier: Thermo Scientific Chemicals
|
Danger
|
L14582-AA
L14582-AC
CAAAAL14582-AAEA
138.25
CAD
CAAAAL14582-AA
CAAAAL14582-AC
CAAAAL14582-XF
(R)-(+)-2-Methyl-CBS-oxazaborolidine 1 M in toluene
(R)-(+)-2-Methyl-CBS-oxazaborolidine
Fieser: 14,239 16,253 17,242
CAS No.: 112022-83-0
Molecular Formula: C18H20BNO
Formula Weight: 277.17
Flash Point: 4°(39°F)
UN No.: 1993
MDL No.: MFCD00078440
Formula:
C₁₈H₂₀BNO Density: 0.925 Flash Pt: 4 °C (39 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00078440 CAS Number: 112022-83-0 UN: 1294 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...