1-Bromo-6-phenylhexane 97%
Supplier: Thermo Scientific Chemicals
L14568-03
L14568-06
CAAAAL14568-03EA
167.48
CAD
CAAAAL14568-03
CAAAAL14568-06
CAAAAL14568-MD
1-Bromo-6-phenylhexane 97%
1-Bromo-6-phenylhexane
Purity: 97%
CAS No.: 27976-27-8
Molecular Formula: C6H5(CH2)6Br
Formula Weight: 241.18
Refractive Index: 1.5295
Boiling Point: 115-118°/6mm
Beilstein Reference: 2207649
MDL No.: MFCD00236022
Formula:
C₆H₅(CH₂)₆Br Boiling Pt: 115…118 °C/6 mm Storage Temperature: Ambient |
MDL Number:
MFCD00236022 CAS Number: 27976-27-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...