4-(2-Hydroxyhexafluoroisopropyl)benzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L14554-03
L14554-06
CAAAAL14554-03EA
161.16
CAD
CAAAAL14554-03
CAAAAL14554-06
4-(2-Hydroxyhexafluoroisopropyl)benzoic acid 97%
4-(2-Hydroxyhexafluoroisopropyl)benzoic acid
Purity: 97%
CAS No.: 16261-80-6
Molecular Formula: C10H6F6O3
Formula Weight: 288.15
Odor: Odorless
Melting Point: 122-126°
Beilstein Reference: 2148278
MDL No.: MFCD00236043
Formula:
C₁₀H₆F₆O₃ Melting Pt: 122…126 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236043 CAS Number: 16261-80-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...