Methyl-3-hydroxy-2-naphthoate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-hydroxy-2-naphthoate

L14533-06 L14533-14
CAAAAL14533-06EA 85.79 CAD
CAAAAL14533-06 CAAAAL14533-14
Methyl-3-hydroxy-2-naphthoate 98+%
Methyl 3-hydroxy-2-naphthoate

Purity: 98+% CAS No.: 883-99-8 Molecular Formula: C12H10O3 Formula Weight: 202.21 Melting Point: 72-74° Boiling Point: 205-207°/160mm MDL No.: MFCD00004099

Formula: C₁₂H₁₀O₃
MW: 202.21 g/mol
Boiling Pt: 205…207 °C/160 mm
Melting Pt: 72…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00004099
CAS Number: 883-99-8
EINECS: 212-936-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR