3',5-Dihydroxy-4',6,7-trimethoxyflavone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-6,7-dimethoxy-4H-chromen-4-one, Eupatorin

L14495-MB L14495-MC
CAAAAL14495-MBEA 478.74 CAD
CAAAAL14495-MB CAAAAL14495-MC
3',5-Dihydroxy-4',6,7-trimethoxyflavone 97%
3',5-Dihydroxy-4',6,7-trimethoxyflavone

Purity: 97% CAS No.: 855-96-9 Molecular Formula: C18H16O7 Formula Weight: 344.32 Melting Point: 190-192° Beilstein Reference: 1355109 MDL No.: MFCD00016929

Formula: C₁₈H₁₆O₇
MW: 344.32 g/mol
Melting Pt: 190…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00016929
CAS Number: 855-96-9
Merck Index: 14,3901

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR