(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

L14422-03 L14422-06 L14422-14
CAAAAL14422-03EA 41.08 CAD
CAAAAL14422-03 CAAAAL14422-06 CAAAAL14422-14
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid 97%
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

Purity: 97% CAS No.: 74163-81-8 Molecular Formula: C10H11NO2 Formula Weight: 177.20 Melting Point: 300° dec. Beilstein Reference: 4842199

Formula: C₁₀H₁₁NO₂
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00144533
CAS Number: 74163-81-8

Order Now

Specification Test Results

Optical rotation -163° (c=2 in 1N NaOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR