Pyromellitic diimide 97%

Supplier: Thermo Scientific Chemicals

L14372-06 L14372-14
CAAAAL14372-06EA 71.26 CAD
CAAAAL14372-06 CAAAAL14372-14
Pyromellitic diimide 97%
Pyromellitic diimide

Purity: 97% CAS No.: 2550-73-4 Molecular Formula: C10H4N2O4 Formula Weight: 216.15 Melting Point: 300° Beilstein Reference: 214780 MDL No.: MFCD00005004

Formula: C₁₀H₄N₂O₄
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00005004
CAS Number: 2550-73-4
EINECS: 219-852-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR