Pentaethylene glycol di-p-toluenesulfonate 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Tos-PEG6-Tos, Pentaethylene glycol di(p-toluenesulphonate), Pentaethylene glycol di(p-toluenesulfonate)

L14369-03 L14369-06
CAAAAL14369-03EA 53.4 CAD
CAAAAL14369-03 CAAAAL14369-06
Pentaethylene glycol di-p-toluenesulfonate 95%
Pentaethylene glycol di-p-toluenesulfonate

Purity: 95% CAS No.: 41024-91-3 Molecular Formula: C24H34O10S2 Formula Weight: 546.66 Flash Point: 110°(230°F) Refractive Index: 1.5225 Beilstein Reference: 3599245 MDL No.: MFCD00012204

Formula: C₂₄H₃₄O₁₀S₂
MW: 546.66 g/mol
Density: 1.260
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012204
CAS Number: 41024-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR