Naphthalene-1,4,5,8-tetracarboxylic dianhydride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: NTCDA, Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride, 1,4,5,8-Naphthalenetetracarboxylic dianhydride

L14365-06 L14365-14
CAAAAL14365-06EA 76.31 CAD
CAAAAL14365-06 CAAAAL14365-14
Naphthalene-1,4,5,8-tetracarboxylic dianhydride 97%
Naphthalene-1,4,5,8-tetracarboxylic dianhydride

Purity: 97% CAS No.: 81-30-1 Molecular Formula: C14H4O6 Formula Weight: 268.18 Melting Point: 300° Beilstein Reference: 272788 MDL No.: MFCD00006915

Formula: C₁₄H₄O₆
MW: 268.18 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006915
CAS Number: 81-30-1
EINECS: 201-342-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR