Methacrylic anhydride 94% stabilised

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methacrylic Anhydride [stabilized with 1,1,3-Tris(3-tert-butyl-4-hydroxy-6-methylphenyl)butane]

Stabilization: ∼ 0.2% Stabilised with 2,4-dimethyl-6-tert-butylphenol

L14357-18 L14357-30
CAAAAL14357-18EA 54.19 CAD
CAAAAL14357-18 CAAAAL14357-30
Methacrylic anhydride 94% stabilised
Methacrylic anhydride

Stab. with ca 0.2% 2,4-dimethyl-6-tert-butylphenol


Purity: 94% CAS No.: 760-93-0 Molecular Formula: C8H10O3 Formula Weight: 154.17 Flash Point: 84°(183°F) Refractive Index: 1.4540 Melting Point: -109° Boiling Point: 197° Beilstein Reference: 1761982 UN No.: 3265 MDL No.: MFCD00008586

Formula: C₈H₁₀O₃
MW: 154.165 g/mol
Boiling Pt: 197 °C
Melting Pt: –109 °C
Density: 1.033
Flash Pt: 84 °C (183 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008586
CAS Number: 760-93-0
EINECS: 212-084-8
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR