Bis(triphenylphosphoranylidene)ammonium chloride, (max. 3% H₂O) 97% (dry weight)
Supplier: Thermo Scientific Chemicals
|
Warning
|
L14339-06
L14339-14
CAAAAL14339-06EA
126.56
CAD
CAAAAL14339-06
CAAAAL14339-14
Bis(triphenylphosphoranylidene)ammonium chloride, (max. 3% H₂O) 97% (dry weight)
Bis(triphenylphosphoranylidene)ammonium chloride
Purity: 97%
CAS No.: 21050-13-5
Molecular Formula: C36H30ClNP2
Formula Weight: 574.05
Melting Point: 274-276°
Beilstein Reference: 4097979
MDL No.: MFCD00151523
Formula:
C₃₆H₃₀ClNP₂ Melting Pt: 274…276 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151523 CAS Number: 21050-13-5 EINECS: 244-170-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...