(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(+)-Mosher's Acid Chloride, (+)-MTPA-Cl, (S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, (S)-3,3,3-Trifluoro-2-methoxy-2-phenylpropanoyl chloride
Purity: 98+%
CAS No.: 20445-33-4
Molecular Formula: C10H8ClF3O2
Formula Weight: 252.62
Storage Temperature: -30°C to -10°C
Flash Point: 89°(192°F)
Refractive Index: 1.4690
Boiling Point: 213-214°
Beilstein Reference: 3591564
Optical Rotation: +137° (c=6.4 in carbon tetrachloride)
UN No.: 3265
MDL No.: MFCD00067105
Formula:
C₁₀H₈ClF₃O₂ Boiling Pt: 213…214 °C Density: 1.353 Flash Pt: 89 °C(192 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00067105 CAS Number: 20445-33-4 UN: 3265 ADR: "8",III Merck Index: 14,6280 |
Specification Test Results
Optical rotation | +137° (c=6.4 in carbon tetrachloride) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...