(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L14325-03
L14325-MD
L14325-MF
CAAAAL14325-03EA
802.64
CAD
CAAAAL14325-03
CAAAAL14325-MD
CAAAAL14325-MF
(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥98%
(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
Purity: 98+%
CAS No.: 39637-99-5
Molecular Formula: C10H8ClF3O2
Formula Weight: 252.62
Storage Temperature: -30°C to -10°C
Flash Point: 89°(192°F)
Refractive Index: 1.4690
Boiling Point: 213-214°
Beilstein Reference: 3591565
Optical Rotation: -137° (neat)
UN No.: 3265
MDL No.: MFCD00044400
Formula:
C₁₀H₈ClF₃O₂ Boiling Pt: 213…214 °C Density: 1.353 Flash Pt: 89 °C(192 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00044400 CAS Number: 39637-99-5 UN: 3265 ADR: "8",III Merck Index: 14,6280 |
Specification Test Results
Optical rotation | -137° (c=6.4 in carbon tetrachloride) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...