1-Fluoropyridiniumtrifluoromethanesulfonate 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-Fluoropyridinium trifluoromethanesulfonate, N-Fluoropyridinium triflate, 1-Fluoropyridiniumtriflate, N-Fluoropyridinium trifluoromethanesulphonate
L14324-03
L14324-MD
CAAAAL14324-03EA
126.72
CAD
CAAAAL14324-03
CAAAAL14324-MD
1-Fluoropyridiniumtrifluoromethanesulfonate 95%
1-Fluoropyridinium trifluoromethanesulphonate
Fieser: 15,160 16,170
Purity: 95%
CAS No.: 107263-95-6
Molecular Formula: C6H5F4NO3S
Formula Weight: 247.17
Melting Point: 180-185°
Beilstein Reference: 4222821
UN No.: 3261
MDL No.: MFCD00013458
Formula:
C₆H₅F₄NO₃S Melting Pt: 180…185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013458 CAS Number: 107263-95-6 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...