Lauroyl peroxide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L14310-22
L14310-36
CAAAAL14310-22EA
97.33
CAD
CAAAAL14310-22
CAAAAL14310-36
Lauroyl peroxide 97%
Lauroyl peroxide
Fieser: 20,147 21,173
Purity: 97%
CAS No.: 105-74-8
Molecular Formula: [CH3(CH2)10CO2]2
Formula Weight: 398.63
Melting Point: 53-57°
Beilstein Reference: 1804936
UN No.: 3106
MDL No.: MFCD00008964
Formula:
(C₁₁H₂₃CO)₂O₂ Melting Pt: 53…57 °C Density: 0.910 Storage Temperature: Ambient |
MDL Number:
MFCD00008964 CAS Number: 105-74-8 EINECS: 203-326-3 UN: 3106 ADR: "5.2", |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...