1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-O-Acetyl-2,3,5-Tri-O-benzoyl-beta-D-Ribofuranose, (2S,3R,4R,5R)-2-Acetoxy-5-((benzoyloxy)methyl)tetrahydrofuran-3,4-diyl dibenzoate, β-D-Ribofuranose 1-Acetate 2,3,5-Tribenzoate, 2,3,5-Tri-o-Benzoyl-1-o-Acetyl-β-D-Ribofuranose, 1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose, β-D-Ribofuranose 1-acetate 2,3,5-tribenzoate

L14302-03 L14302-06
CAAAAL14302-03EA 48.03 CAD
CAAAAL14302-03 CAAAAL14302-06
1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose 98%
1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose

Purity: 98% CAS No.: 6974-32-9 Molecular Formula: C28H24O9 Formula Weight: 504.50 Melting Point: 128-130° Beilstein Reference: 100243 Optical Rotation: +44° (c=0.5 in chloroform) MDL No.: MFCD00005357

Formula: C₂₈H₂₄O₉
MW: 504.5 g/mol
Melting Pt: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00005357
CAS Number: 6974-32-9
EINECS: 230-220-4

Order Now

Specification Test Results

Optical rotation +44° (c=0.5 in chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR