Amberlite® IR-120 (H) ion exchange resin

Supplier: Thermo Scientific Chemicals
Danger

L14285-0B L14285-30
CAAAAL14285-0BEA 145.52 CAD
CAAAAL14285-0B CAAAAL14285-30
Amberlite® IR-120 (H) ion exchange resin
Amberlite® IR-120 (H)

Fieser: 1,511 4,266 5,355 6,302 9,256


CAS No.: 78922-04-0 MDL No.: MFCD00132707

Density: 1.28
Storage Temperature: Ambient
MDL Number: MFCD00132707
CAS Number: 78922-04-0
Merck Index: 14,382

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR