Dowex® 1X8 (Cl), Dowex® 200-400 mesh

Supplier: Thermo Scientific Chemicals

L14258-0B L14258-18 L14258-30
CAAAAL14258-0BEA 894.28 CAD
CAAAAL14258-0B CAAAAL14258-18 CAAAAL14258-30
Dowex® 1X8 (Cl), Dowex® 200-400 mesh
Dowex® 1X8 (Cl)

CAS No.: 12627-85-9 Odor: Amine-like MDL No.: MFCD00132718

Storage Temperature: Ambient
MDL Number: MFCD00132718
CAS Number: 12627-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR