Dowex® 1X8 (Cl), Dowex® 100-200 mesh
Supplier: Thermo Scientific Chemicals
L14257-0B
L14257-18
L14257-30
CAAAAL14257-0BEA
933.78
CAD
CAAAAL14257-0B
CAAAAL14257-18
CAAAAL14257-30
Dowex® 1X8 (Cl), Dowex® 100-200 mesh
Dowex® 1X8 (Cl)
CAS No.: 12627-85-9
Odor: Amine-like
MDL No.: MFCD00132718
Storage Temperature:
Ambient |
MDL Number:
MFCD00132718 CAS Number: 12627-85-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...