Dowex® 1X2 (Cl), Dowex® 100-200 mesh

Supplier: Thermo Scientific Chemicals

L14254-0B L14254-18 L14254-30
CAAAAL14254-0BEA 1091.78 CAD
CAAAAL14254-0B CAAAAL14254-18 CAAAAL14254-30
Dowex® 1X2 (Cl), Dowex® 100-200 mesh
Dowex® 1X2 (Cl)

CAS No.: 9085-42-1 Odor: Amine-like MDL No.: MFCD00132715

Storage Temperature: Ambient
MDL Number: MFCD00132715
CAS Number: 9085-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR