S-(2-Aminoethyl)isothiouronium bromide hydrobromide 98%
Supplier: Thermo Scientific Chemicals
L14232-06
L14232-14
CAAAAL14232-06EA
48.51
CAD
CAAAAL14232-06
CAAAAL14232-14
S-(2-Aminoethyl)isothiouronium bromide hydrobromide 98%
S-(2-Aminoethyl)isothiouronium bromide hydrobromide
Purity: 98%
CAS No.: 56-10-0
Molecular Formula: C3H9N3S.2HBr
Formula Weight: 281.02
Odor: Odorless
Melting Point: 190-192°
Beilstein Reference: 3911163
MDL No.: MFCD00037011
Formula:
C₃H₁₁Br₂N₃S Melting Pt: 190…192 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037011 CAS Number: 56-10-0 EINECS: 200-257-0 Merck Index: 14,178 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...