9-Bromo-9-phenylfluorene 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L14225-03
L14225-06
CAAAAL14225-03EA
77.74
CAD
CAAAAL14225-03
CAAAAL14225-06
9-Bromo-9-phenylfluorene 96%
9-Bromo-9-phenylfluorene
Fieser: 13,48 15,49 15,49
Purity: 96%
CAS No.: 55135-66-5
Molecular Formula: C19H13Br
Formula Weight: 321.23
Melting Point: 94-99°
Beilstein Reference: 2561445
UN No.: 3261
MDL No.: MFCD00075522
Formula:
C₁₉H₁₃Br Melting Pt: 94…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075522 CAS Number: 55135-66-5 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...