Fisetin 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,7,3',4'-Tetrahydroxyflavone

L14207-03 L14207-MD
CAAAAL14207-03EA 306.52 CAD
CAAAAL14207-03 CAAAAL14207-MD
Fisetin 96%
Fisetin

Purity: 97% CAS No.: 528-48-3 Molecular Formula: C15H10O6 Formula Weight: 286.24 Melting Point: 330° dec. Beilstein Reference: 292829 MDL No.: MFCD00006829

Formula: C₁₅H₁₀O₆
MW: 286.24 g/mol
Melting Pt: 330 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006829
CAS Number: 528-48-3
EINECS: 208-434-4
Merck Index: 14,4088

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR