(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-4-Hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide, (R)-(-)-1,1'-Binaphthalene-2,2'-diyl-phosphate

L14149-03 L14149-MD
CAAAAL14149-03EA 205.4 CAD
CAAAAL14149-03 CAAAAL14149-MD
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate 98+%
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate

Fieser: 12,49 16,25


Purity: 98+% CAS No.: 39648-67-4 Molecular Formula: C20H13O4P Formula Weight: 348.29 Melting Point: 300° Beilstein Reference: 4713363

Formula: C₂₀H₁₃O₄P
MW: 348.289 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00010045
CAS Number: 39648-67-4
UN: 3464
ADR: "6.1",III

Order Now

Specification Test Results

Optical rotation -605° (c=1 in methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR