D-tert-Leucine (H-D-Tle-OH) 99%

Supplier: Thermo Scientific Chemicals

L14098-04 L14098-ME
CAAAAL14098-04EA 919.56 CAD
CAAAAL14098-04 CAAAAL14098-MC CAAAAL14098-ME
D-tert-Leucine (H-D-Tle-OH) 99%
D-tert-Leucine (H-D-Tle-OH)

Purity: 99% CAS No.: 26782-71-8 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Melting Point: 300° Beilstein Reference: 1721825

Formula: C₆H₁₃NO₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004265
CAS Number: 26782-71-8

Order Now

Specification Test Results

Optical rotation -7° (c=5 in 5N HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR