(-)-Menthyl acetate 98%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
L-Menthyl acetate, L-Menthol acetate, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl acetate, (R)-(-)-Menthyl acetate, (-)-Acetoxy-p-menthane, (5R)-2-Isopropyl-5-methylcyclohexyl acetate, (1R)-(−)-Menthyl acetate
L14095-03
L14095-MD
CAAAAL14095-03EA
127.03
CAD
CAAAAL14095-03
CAAAAL14095-MD
(-)-Menthyl acetate 98%
(-)-Menthyl acetate
Purity: 98%
CAS No.: 2623-23-6
Molecular Formula: C12H22O2
Formula Weight: 198.31
Flash Point: 77°(170°F)
Refractive Index: 1.4470
Boiling Point: 229-231°
Beilstein Reference: 2208505
Optical Rotation: -80.5° (c=8 in benzene)
MDL No.: MFCD00001482
Formula:
C₁₂H₂₂O₂ Boiling Pt: 229…231 °C Density: 0.925 Flash Pt: 77 °C(170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001482 CAS Number: 2623-23-6 EINECS: 220-076-0 Merck Index: 14,5839 |
Specification Test Results
Optical rotation | -80.5° (c=8 in benzene) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...