2,3-Dihydro-1H-inden-2-amine hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L14083-03
L14083-06
CAAAAL14083-03EA
70.63
CAD
CAAAAL14083-03
CAAAAL14083-06
CAAAAL14083-14
2,3-Dihydro-1H-inden-2-amine hydrochloride 98%
2,3-Dihydro-1H-inden-2-amine hydrochloride
Purity: 98%
CAS No.: 2338-18-3
Molecular Formula: C9H11N.HCl
Formula Weight: 169.66
Odor: Odorless
Melting Point: 243-248°
Beilstein Reference: 3913700
MDL No.: MFCD00012549
Formula:
C₉H₁₂ClN Melting Pt: 243…248 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012549 CAS Number: 2338-18-3 EINECS: 219-048-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...