2-Amino-4-(4-chlorophenyl)thiazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(4-Chlorophenyl)thiazol-2-amine
L14067-03
L14067-06
CAAAAL14067-03EA
46.93
CAD
CAAAAL14067-03
CAAAAL14067-06
2-Amino-4-(4-chlorophenyl)thiazole 98%
2-Amino-4-(4-chlorophenyl)thiazole
Purity: 98%
CAS No.: 2103-99-3
Molecular Formula: C9H7ClN2S
Formula Weight: 210.69
Melting Point: 168-170°
Beilstein Reference: 153889
Formula:
C₉H₇ClN₂S MW: 210.69 g/mol Melting Pt: 168…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047058 CAS Number: 2103-99-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...