Benzyl (S)-(+)-glycidyl ether 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-2-((Benzyloxy)methyl)oxirane

L14033-03 L14033-06
CAAAAL14033-03EA 63.36 CAD
CAAAAL14033-03 CAAAAL14033-06
Benzyl (S)-(+)-glycidyl ether 98+%
Benzyl (S)-(+)-glycidyl ether

Purity: 98+% CAS No.: 16495-13-9 Molecular Formula: C10H12O2 Formula Weight: 164.20 Flash Point: 110°(230°F) Refractive Index: 1.5170 Boiling Point: 130°/0.1mm Beilstein Reference: 5246495 Optical Rotation: +5.1° (c=5 in toluene) MDL No.: MFCD00054428

Formula: C₁₀H₁₂O₂
MW: 164.20 g/mol
Boiling Pt: 130 °C/0.1 mm
Density: 1.072
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00054428
CAS Number: 16495-13-9

Order Now

Specification Test Results

Optical rotation +5.1° (c=5 in toluene)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR