Sodium-3-chloro-2-hydroxypropanesulfonate hemihydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

L14018-22 L14018-36
CAAAAL14018-22EA 46.45 CAD
CAAAAL14018-22 CAAAAL14018-36
Sodium-3-chloro-2-hydroxypropanesulfonate hemihydrate 98%
Sodium-3-chloro-2-hydroxypropanesulfonate hemihydrate

Purity: 98% CAS No.: 143218-48-8 Molecular Formula: C3H6ClNaO4S.0.5H2O Formula Weight: 205.60 (196.59anhy) Melting Point: ca 256° dec. Beilstein Reference: 4164376 MDL No.: MFCD00149699

Formula: C₃H₆ClNaO₄S·½H₂O
MW: 205.60 g/mol (196.59 anhy)
Melting Pt: ca 256 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149699
CAS Number: 143218-48-8
EINECS: 204-807-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR