Ethyl-5-fluoro-1H-indole-2-carboxylate 98%
Supplier: Thermo Scientific Chemicals
L14010-03
L14010-06
CAAAAL14010-03EA
57.35
CAD
CAAAAL14010-03
CAAAAL14010-06
Ethyl-5-fluoro-1H-indole-2-carboxylate 98%
Ethyl-5-fluoro-1H-indole-2-carboxylate
Purity: 98%
CAS No.: 348-36-7
Molecular Formula: C11H10FNO2
Formula Weight: 207.21
Melting Point: 147-152°
Beilstein Reference: 170254
MDL No.: MFCD00152076
Formula:
C₁₁H₁₀FNO₂ Melting Pt: 147…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00152076 CAS Number: 348-36-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...